Learn More
Invitrogen™ F4/80 Monoclonal Antibody (BM8), Alexa Fluor™ 647

Description
F4/80 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | F4/80 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | BM8 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | PBS with 4mg/mL BSA and 0.1% sodium azide |
| Gene | ADGRE1 |
| Gene Accession No. | Q61549 |
| Gene Alias | ADGRE1; Adhesion G protein-coupled receptor E1; Adhesion G protein-coupled receptor E2; cell surface glycoprotein EMR1; cell surface glycoprotein F4/80; DD7A5-7; EGF-like module containing, mucin-like, hormone receptor-like 1; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-TM7; Emr1; EMR1 hormone receptor; F4/80; Gpf480; Ly71; lymphocyte antigen 71; TM7LN3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.