Learn More
Eppendorf 3P CryoCube F740h, 740 L, classic interface, VIP, green cooling liquids, air cooling, handle left side, 3 shelves, 120 – 208 V/60 Hz (US)
Supplier:  Eppendorf 3P F740540015
Encompass_Preferred
- Traditionally foamed polyurethane insulation enforced by vacuum insulation panels in freezer body provides efficient insulation protection- Quick pull-down and recovery times with reliable heavy-duty compressors for sample safety- Left- or right-handed outer door as option for optimal space usage in the lab- Easy-to-read, eye level, flush mounted LED control panel and alarm status- Inner doors are insulated and sealed, minimizing cold air loss while accessing your samples- 3 or 5 compartments as option, each accepts Eppendorf racks as well as other popular brands without adjustment- Each shelf has air slits for fast temperature adaption- Special low-temperature outer door seals allow gaskets to remain flexible at -86 °C and prevents ice buildup- Full stainless steel interior for easy cleaning during maintenance and defrost cycles; powder-coated steel exterior
Catalog No. NC1895708
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.