Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Series 815 Laser Slotted Cassettes with Attached Lids

Catalog No. 8151018LM Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
81-510-18LM Beige
8150318LM Blue
8150618LM Orange
8150718LM Gray
8150818LM Aqua
8150918LM Lilac
8151218LM Red
81-511-18LM Ochre
8150118LM White
8150218LM Yellow
8150418LM Pink
8150518LM Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 81-510-18LM Supplier Epredia™ Supplier No. 8151018LM
Add to Cart
Add to Cart
This item is not returnable. View return policy

Manufactured specifically for laser printing, the Epredia™ Series 815 Laser Slotted Cassettes come with attached lids and are available in multiple colors.

  • Epredia™ Laser Cassettes are designed for use on both the NOVA™ and VEGA™ Laser Printers
  • Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Each style comes in multiple different colors
  • Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
  • Series 815 come in taped stacks of 40 cassettes with lids attached; 45 stacks per case for a total of 1800 cassettes
  • The Epredia™ Laser Cassette is not compatible with thermal tape style printers

Specifications

Pore Style Slots
Quantity 40 Cassettes/Taped Stack, 45 Stacks/Cs.
Closure Included Yes
Closure Type Attached Lid
Color Beige
Product Type Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.