Learn More
Epredia™ Series 110 Laser Slotted with Front Tab Cassettes with Lids
Catalog No. 1101110LM Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11-011-10LM Ochre
1100210LM Yellow
1100810LM Aqua
1100110LM White
1100310LM Blue
1100410LM Pink
1100510LM Green
1100610LM Orange
1100710LM Gray
1100910LM Lilac
1101210LM Red
11-010-10LM Beige
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 11-011-10LM Supplier Epredia™ Supplier No. 1101110LM
Add to Cart
Add to Cart
This item is not returnable. View return policy
Manufactured specifically for laser printing, the Epredia™ Series 110 Slotted Cassettes come with separate lids with a tab in the front for easy opening and closing.
  • Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
  • Contain a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Each style comes in different colors with color-matched lids
  • Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
  • Comes in taped stacks of 40 cassettes; 25 stacks per case (for a total of 1000 cassettes with separate lids)
  • The Epredia™ Laser Cassette is not compatible with thermal tape style printers
Specifications
Pore Style Slots
Quantity 40 Cassettes/Taped Stack, 25 Stacks/Cs.
Closure Included Yes
Closure Type Separate Lid
Color Ochre
Product Type Cassette