Description
- Epredia™ Laser Cassettes are designed for use on both the NOVA™ and VEGA™ Laser Printers.
- They contain a special constituent chemical compound that carbonizes when exposed to the laser beam.
- Each style comes in 10 different colors: white, yellow, blue, pink, green, orange, grey, aqua, lilac and red with color-matched lids.
- Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing.
- Series 110, 115 and 215 come in taped stacks of 40 cassettes; 25 stacks per case.
- Series 815 come in taped stacks of 40 cassettes with lids attached; 45 stacks per case.
- The Epredia™ Laser Cassette is not compatible with thermal tape style printers.
Specifications
Specifications
| Pore Style | Small Squares |
| Quantity | 40 Cassettes/Taped Stack, 25 Stacks/Cs. |
| Closure Included | Yes |
| Closure Type | Separate Lid |
| Color | Red |
| Product Type | Cassette |