Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Series 315 Laser Embedding Cassettes

Catalog No. 3150218LM Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
3150218LM Yellow
3150118LM White
3150318LM Blue
3150418LM Pink
3150518LM Green
3150618LM Orange
3150718LM Gray
3150818LM Aqua
3150918LM Lilac
3151218LM Red
31-510-18LM Beige
31-511-18LM Ochre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 3150218LM Supplier Epredia™ Supplier No. 3150218LM
Add to Cart
Add to Cart
This item is not returnable. View return policy

Manufactured specifically for laser printing, the Epredia™ Series 315 Embedding Cassettes have round holes to maximize fluid exchange.

  • Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
  • Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Embedding style of cassette comes in different colors
  • Required reusable metal cassette lids (Part number FTML315) are sold separately
  • Embedding cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
  • Series 315 comes in taped stacks of 40 cassettes; 25 stacks per case
  • Epredia™ Laser Cassette Series is not compatible with thermal tape style printers

Specifications

Pore Style Round Holes
Quantity 40 Cassettes/Taped Stack, 45 Stacks/Cs.
Closure Included No
Color Yellow
Product Type Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.