Learn More
Description
- Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
- Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
- Embedding style of cassette comes in different colors
- Required reusable metal cassette lids (Part number FTML315) are sold separately
- Embedding cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
- Series 315 comes in taped stacks of 40 cassettes; 25 stacks per case
- Epredia™ Laser Cassette Series is not compatible with thermal tape style printers
Specifications
Specifications
| Pore Style | Round Holes |
| Quantity | 40 Cassettes/Taped Stack, 45 Stacks/Cs. |
| Closure Included | No |
| Color | Red |
| Product Type | Cassette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.