Learn More
Description
- Epredia™ Laser Cassettes are designed for use on both the NOVA™ and VEGA™ Laser Printers
- Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
- Series 615 comes in multiple different colors with color-matched lids
- Designed to maximize fluid exchange during processing while still protecting very small and friable tissue samples without the use of sponges or lens paper
- Come in stacks stripped together by string for ease of loading into Epredia™ cassette hoppers for printing
- Series 615 comes in stripped stacks of 40 cassettes; 25 stacks per case
- Epredia™ Laser Cassette is not compatible with thermal tape style printers
Specifications
Specifications
| Pore Style | Very Small Squares |
| Quantity | 40 Cassettes/Taped Stack, 25 Stacks/Cs. |
| Closure Included | Yes |
| Closure Type | Separate Lid |
| Color | Aqua |
| Product Type | Cassette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.