Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAAH Recombinant Rabbit Monoclonal Antibody (10A1M7)
GREENER_CHOICE

Catalog No. PIMA535442
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535442 Supplier Invitrogen™ Supplier No. MA535442
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DLNAPGRATG AVSYTMLYNC LDFPAGVVPV TTVTAEDEAQ MEHYRGYFGD IWDKMLQKGM KKSVGLPVAV QCVALPWQEE LCLRFMREVE RLMTPEKQSS.

Degrades bioactive fatty acid amides like oleamide, the endogenous levels of cannabinoid, anandamide and myristic amide to their corresponding acids, thereby serving to terminate the signaling functions of these molecules. Hydrolyzes polyunsaturated substrate anandamide preferentially as compared to monounsaturated substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAAH
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 10A1M7
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene FAAH
Gene Accession No. O00519, O08914, P97612
Gene Alias amide hydrolase; anandamide amidohydrolase 1; AW412498; FA2H; Faah; FAAH1; FAAH-1; fatty acid amide hydrolase; fatty acid amide hydrolase protein; fatty-acid amide hydrolase 1; fatty-acid amide hydrolase 1-like; LOC100911581; MGC102823; MGC138146; Oleamide hydrolase; Oleamide hydrolase 1; PSAB
Gene Symbols FAAH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 480-579 of human FAAH1 (O00519).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14073, 2166, 29347
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.