Learn More
Rockland Immunochemicals Fab Mouse IgG (H&L) Antibody Peroxidase Conjugated 1 mg
SDP
Supplier:  Rockland Immunochemicals 8101302
Encompass
Fab Anti-Mouse IgG (H&L) Peroxidase Antibody generated in goat detects Mouse IgG. This product possesses the F(ab) region possessing the epitope-recognition site both heavy and light chains of the antibody molecule are present. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3958
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.