Learn More
Sartorius Octet Anti-Human Fab-CH1 Biosensors, High-Affinity Ligand, Enhanced Baseline Stability, Tray of 96
Catalog No. NC0936561 Shop All Sartorius Products
Encompass
The Octet Anti-Human Fab-CH1 2nd Generation (FAB2G) Biosensors provide precise and efficient analysis of human Fab fragments and full-length IgG through Bio-Layer Interferometry technology. These biosensors ensure oriented capture with minimal steric hindrance, allowing for label-free and rapid analysis from complex samples. Enhanced baseline stability and regeneration capabilities differentiate FAB2G from previous models, making it an essential tool for applications in lead identification and QC in drug research.
Catalog No. NC0936561 Supplier Sartorius Supplier No. 185125
The Octet Anti-Human Fab-CH1 2nd Generation (FAB2G) Biosensors provide precise and efficient analysis of human Fab fragments and full-length IgG through Bio-Layer Interferometry technology. These biosensors ensure oriented capture with minimal steric hindrance, allowing for label-free and rapid analysis from complex samples. Enhanced baseline stability and regeneration capabilities differentiate FAB2G from previous models, making it an essential tool for applications in lead identification and QC in drug research.