Learn More
Advanced Targeting Systems FABFC-PHAST HUMAN PH-10 250UG
Supplier:  Advanced Targeting Systems PH10250
Encompass
FabFc-pHast human is one of our fastest tools for quantitative testing of your primary antibody’s specificity, binding, and internalization, providing results in 1 day. FabFc-pHast human binds to your primary human antibody via a secondary antibody cross-linked to a pH-dependent fluorescent reporter. This fluorescent reporter will increase intensity as the pH of its surroundings becomes more acidic, as evident when exposed to the environment inside a cell. A successful assay will provide an EC50 by way of a fluorescence detecting plate reader, illuminating your lead antibody candidates. The FabFc-pHast products can be used in B-cell research with the confidence that fluorescence is primary antibody-mediated.
Catalog No. NC2596022
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.