Learn More
Sartorius Incucyte Human Fabfluor-pH Antibody Labeling Dye for Antibody Internalization, Orange, 50 ug
Catalog No. NC1841090 Shop All Sartorius Products
Encompass
One vial (50 ug) of lyophilized conjugate sufficient for labeling large quantities of human test antibodies in 96-well format.
Catalog No. NC1841090 Supplier Sartorius Supplier No. 4812
One vial (50 ug) of lyophilized conjugate sufficient for labeling large quantities of human test antibodies in 96-well format.