Learn More
Sartorius Incucyte Human Fabfluor-pH Antibody Labeling Dye for Antibody Internalization
Catalog No. NC1417770 Shop All Sartorius Products
Encompass
One vial (50 ug) of lyophilized conjugate sufficient for labeling large quantities of human test antibodies in 96-well format.
Catalog No. NC1417770 Supplier Sartorius Supplier No. 4722
One vial (50 ug) of lyophilized conjugate sufficient for labeling large quantities of human test antibodies in 96-well format.