Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7205155 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8887 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8887 Supplier Proteintech Group Inc Supplier No. 136261AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This protein binds free fatty acids and their coenzyme a derivatives, bilirubin, and some other small molecules in the cytoplasm; it may be involved in intracellular lipid transport. Also seems to bind selenium.

Specifications

Antigen FABP1
Applications Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FABP1
Gene Accession No. P02692, P07148, P12710
Gene Alias FABP1, FABPL, Fatty acid binding protein 1, L FABP
Gene Symbols FABP1
Host Species Rabbit
Immunogen FABP1 Fusion Protein Ag4540
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14080, 2168, 24360
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.