Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FABP3 (Human) ELISA Kit

Catalog No. 89032271 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-271 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-271 Supplier Abnova™ Supplier No. KA4903
Add to Cart
Add to Cart

Sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human FABP3

  • For quantitative detection of human HFABP in serum, cell culture supernatants or tissue homogenate

Specifications

Calibration Range 15.6 to 1000 pg/mL
Description FABP3 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human FABP3.
Conjugate HRP
Detection Method Colorimetric
Product Type FABP3 (Human) ELISA Kit
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates
Target Species Human
For Use With (Application) Quantification
Gene ID (Entrez) 2170
Gene Symbol FABP3
Kit Contents One 96-well plate pre-coated with anti-human HFABP antibody: 96 (8x12) wells, Lyophilized human HFABP standards: 10ng/tube x 2;
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL (Prediluted)
Storage Requirements Store at 4°C for 4 months, or at -20°C for 8 months. Store HRP conjugated antibody at 2-8°C.
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.