Learn More
Grainger Face Mill FMAL2500903 FM-AL 2.5000 Dia
Supplier:  Grainger 170KA4
Encompass_Preferred
The Sandvik Coromant facemill cutter is a milling cutter used for facing operations. It has a diameter of 2 inches and an overall length of 3 inches. The cutter features 5 indexable carbide inserts with the CoroTurn 107 grade for steel machining applications. It has a right-hand spiral flute design and is balanced to 25,000 RPM. The cutter shank has a Coromant Capto C5 interface. This facemill is well-suited for facing, shoulder milling and slotting applications in steel.
Catalog No. 50-276-4245