Learn More
Jackson Safety Face Shield Cap Adapter
The Jackson Safety 282 B Cord Mounted Cap Adapters for Non-Slotted Caps features a polyethylene spark deflector and is ideal for when the job requires a hard hat and a face shield.
Supplier:  Jackson Safety 14948
 View more versions of this product
Catalog No. 19-060-790
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Comes in 3 Models: Model 37, 137, and 337
  • Curved injection molded channel fit over the hard hat brim (Model 337)
  • Features adjustable rubber strap headband that stretches around the cap crown for a secure fit (Model 337)
  • Features five easy-to-swivel lock mounting pins for universal windows
  • Vertical lock-in eliminates face shield creep (Model 37, 137)
  • Curved metal channels fit over the hard hat brim
  • Brackets feature durable expandable spring tensioner (Model 37, 137)Features five easy-to-swivel lock mounting pins for universal windows
Specifications
Specifications
Silver
Spring
Regular Brim
Regular Brim Hard Hat, Universal Faceshields
Universal
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.