Learn More
Honeywell™ Face Shield Systems
Catalog No. 19017864 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
F-400 headgear
Catalog No.
19-017864 F-400 headgear
1 options
This item is not returnable. View return policy
Catalog No. 19-017864 Supplier Honeywell™ Supplier No. F400
  • $63.75
    $43.65 / Each of 1
    Save $20.10
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Stronger, more durable and more heat-resistant than ordinary face shields.
  • Made from Noryl™, the same material as used in Honeywell welding helmets
  • Includes 4 in. crown only
  • Visor not included
Order Info
Note: Visor is sold separately. (See Cat. No. 19-017-927.)
Specifications
Certifications/Compliance Meets ANSI; CSA; SEI certified
Description 4 in. Crown