Learn More
Medchemexpress LLC Factor D inhibitor 6 | 1386455-51-1 | 99.2% | 100 MG

Supplier: Medchemexpress LLC HY122700100MG
Factor D inhibitor 6 is a potent, selective, and orally active small-molecule inhibitor of complement Factor D (FD) supplied as a white to off-white powder for research use.
- Potent FD inhibition: IC50 30 nM, Kd 6 nM.
- Target: complement Factor D; useful for complement pathway studies.
- High purity: 99.2% (HPLC).
- Form: white to off-white solid (powder), soluble in DMSO.
- Storage: powder -20°C for long-term stability; in solvent store cold (see SDS).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.