Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579034 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579034 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579034 Supplier Invitrogen™ Supplier No. PA579034
  • $525.00
    $429.65 / Each of 1

    Save $95.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human THP-1 whole cell, human Hela whole cell, rat lung tissue, mouse lung tissue, mouse NIH/3T3 whole cell. IHC: human pancreatic cancer tissue, human breast cancer tissue, mouse adipose tissue, rat kidney tissue.

Complement factor D is a serine protease of the alternative pathway of complement activation. The protein is a single chain polypeptide with a molecular mass of 25 kDa. Factor D cleaves factor B bound to C3b, generating the alternative pathway C3 convertase C3bBb and releasing the Ba fragment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor D
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene CFD
Gene Accession No. P00746, P03953, P32038
Gene Alias 28 kDa adipocyte protein; adipsin; Adn; C3 convertase activator; Cfd; Complement factor D; complement factor D (adipsin); complement factor D preproprotein; D component (adipsin) of complement; D component of complement (adipsin); Df; EC 3.4.21.46; Endogenous vascular elastase; EVE; PFD; Properdin factor D
Gene Symbols CFD
Host Species Rabbit
Immunogen E. coli-derived human Factor D recombinant protein (Position: I26-A253).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11537, 1675, 54249
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.