Learn More
Siemens Healthineers Factor II Deficient Plasma Reagent
Catalog No. 23044687 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
3 x 1 mL
Catalog No. Quantity
23-044-687 3 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-687 Supplier Siemens Healthineers Supplier No. 10446330
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the determination of the activity of coagulation factor II (prothrombin) in human plasma by coagulometric methods.
Factor II Deficient Plasma is a human plasma-based reagent for the detection of hereditary or acquired deficiencies of factor II (prothrombin). It is manufactured by immunoadsorption and contains a residual factor concentration of <1% prothrombin and normal levels of fibrinogen and other clotting factors. Factor II Deficient Plasma was designed to be used in combination with Dade Innovin or Thromborel S reagents.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Single Factor Testing
Quantity 3 x 1 mL
WARNING: Cancer - www.P65Warnings.ca.gov