Learn More
Siemens Healthineers Factor V Deficient Plasma Reagent
Catalog No. 23044684 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
8 x 1 mL
Catalog No. Quantity
23-044-684 8 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-684 Supplier Siemens Healthineers Supplier No. 10446271
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the determination of the activity of coagulation factor V in human plasma by coagulometric methods.
Factor V Deficient Plasma is a human plasma-based reagent for the detection of hereditary or acquired deficiencies of factor V. It is manufactured by immunoadsorption and contains a residual factor concentration of <1% factor V and normal levels of fibrinogen and other clotting factors. Factor V Deficient Plasma was designed to be used in combination with Dade Innovin or Thromborel S reagents.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Single Factor Testing
Quantity 8 x 1 mL
WARNING: Cancer - www.P65Warnings.ca.gov