Learn More
Siemens Healthineers Factor V Leiden Reagent
Catalog No. 23044724 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
5 x 4 mL
Catalog No. Quantity
23-044-724 5 x 4 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-724 Supplier Siemens Healthineers Supplier No. 10459420
Add to Cart
Add to Cart
This item is not returnable. View return policy
Russell Viper Venom-based test for screening of resistance to Activated Protein C (APC) in plasma from individuals with the Factor V (Leiden) defect.
The Factor V Leiden assay is a simple functional clotting test system intended for screening of resistance to activated protein C (APC) in plasma from individuals with the factor V Leiden defect. Siemen's Factor V Leiden assay is based on the activation of endogenous protein C by incubation of plasma with Agkistrodon contortrix contortrix (southern copperhead) venom. A dilute Russell’s viper venom time (DRVVT) test is then performed on the plasma.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Thrombophilia Testing
Quantity 5 x 4 mL