Learn More
Siemens Healthineers Factor VII Deficient Plasma Reagent
Catalog No. 23044688 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
3 x 1 mL
Catalog No. Quantity
23-044-688 3 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-688 Supplier Siemens Healthineers Supplier No. 10446407
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the determination of the activity of coagulation factor VII in human plasma by coagulometric methods.
Factor VII Deficient Plasma is a human plasma-based reagent for the detection of hereditary or acquired deficiencies of factor VII. It is manufactured by immunoadsorption and contains a residual factor concentration of <1% factor VII and normal levels of fibrinogen and other clotting factors. Factor VII Deficient Plasma was designed to be used in combination with Dade Innovin or Thromborel S reagents.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Single Factor Testing
Quantity 3 x 1 mL
WARNING: Cancer - www.P65Warnings.ca.gov