Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor VII Monoclonal Antibody (9031)
GREENER_CHOICE

Catalog No. 01676484
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-484 Supplier Invitrogen™ Supplier No. MA143038
  • $599.00
    $497.30 / Each of 1

    Save $101.70

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MA1-43038 detects Factor VII in mouse samples. MA1-43038 has successfully been used in Western blot and ELISA applications. MA1-43038 was derived from mouse factor VII.

Coagulation factor VII is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Defects in F7 production can cause coagulopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor VII
Applications ELISA, Western Blot
Classification Monoclonal
Clone 9031
Conjugate Unconjugated
Formulation 50% water with 50% glycerol and no preservative
Gene F7
Gene Accession No. P70375
Gene Alias AI132620; Cf7; Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; F7; Factor 7; Factor VII heavy chain; Factor VII light chain; Factor7; FVII; FVII coagulation protein; proconvertin; Serum prothrombin conversion accelerator; SPCA
Gene Symbols F7
Host Species Rat
Immunogen Mouse Factor VII.
Purification Method SDS-PAGE
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14068
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.