Learn More
Siemens Healthineers Factor VIII Chromogenic Kit
Catalog No. 23044660 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
2 x 3 mL
Catalog No. Quantity
23-044-660 2 x 3 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-660 Supplier Siemens Healthineers Supplier No. 10445729
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determination of Factor VIII (antihemophilic factor) activity in human plasma.
Factor VIII Chromogenic Reagent is used for the determination of factor VIII concentration in plasma preparations and the detection of hereditary or acquired factor VIII deficiencies. The chromogenic method is recommended for therapeutic factor VIII preparations and is insensitive to heparin at levels of <10 U/mL.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Single Factor Testing
Quantity 2 x 3 mL