Learn More
Siemens Healthineers Factor VIII Deficient Plasma Reagent
Catalog No. 23044689 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
8 x 1 mL
Catalog No. Quantity
23-044-689 8 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-689 Supplier Siemens Healthineers Supplier No. 10446411
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the determination of the activity of coagulation factor VIII in human plasma by coagulometric methods.
Factor VIII Deficient Plasma is a human plasma-based reagent for the detection of hereditary or acquired deficiencies of factor VIII. It is manufactured by immunoadsorption and contains a residual factor concentration of <1% factor VIII and normal levels of fibrinogen and other clotting factors. Factor VIII Deficient Plasma was designed to be used in combination with Dade Actin, Dade Actin FS, Dade Actin FSL, or Pathromtin SL reagents.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Single Factor Testing
Quantity 8 x 1 mL