Learn More
Siemens Healthineers Factor X Deficient Plasma Reagent
Catalog No. 23044691 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
3 x 1 mL
Catalog No. Quantity
23-044-691 3 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-691 Supplier Siemens Healthineers Supplier No. 10446415
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the determination of the activity of coagulation factor X in human plasma by coagulometric methods.
Factor X Deficient Plasma is a human plasma-based reagent for the detection of hereditary or acquired deficiencies of factor X. It is manufactured by immunoadsorption and contains a residual factor concentration of <1% factor X and normal levels of fibrinogen and other clotting factors. Factor X Deficient Plasma was designed to be used in combination with Dade Innovin or Thromborel S reagents.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Single Factor Testing
Quantity 3 x 1 mL