Learn More
Abcam Factor X Human Chromogenic Activity Assay Kit, 100 Tests
Description
- Assay Type: Enzyme activity
- Detection Method: Colorimetric
- Range: 0.016 to 4 μg/mL
- Sensitivity: 0.009 μg/mL
- Sample Types: Plasma, Cell culture supernatant
- Species Reactivity: Human
- Results Type: Quantitative
- Applications: Enzyme activity assay
- Citation Count: 1
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Factor X (Human) |
| Reactivity | Human |
| For Use With (Equipment) | Microplate Reader |
| No. of Reactions | 100 Tests |
| Format | Kit |
| Concentration | 0.016 to 4 μg/mL |
| Detection Location | In-Solution Detection |
| Detection Method | Colorimetric |
| Cross Reactivity | Human |
| Sample Type | Plasma, Cell culture supernatant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.