Learn More
Haematologic Technologies Inc Coagulation Factor Xa, 1 mg
Supplier:  Haematologic Technologies Inc HCXA00601MG
Encompass_Preferred
Factor Xa is prepared by activating purified factor X with the factor X activator isolated from Russell's viper venom. Factor Xa is purified from the activation mixture by chromatography over benzamidine-Sepharose followed by gel filtration. Several modified forms of factor Xa are also available.
Catalog No. NC0732627
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.