Learn More
Siemens Healthineers Factor XI Deficient Plasma Reagent
Catalog No. 23044685 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
3 x 1 mL
Catalog No. Quantity
23-044-685 3 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-685 Supplier Siemens Healthineers Supplier No. 10446316
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the determination of the activity of coagulation factor XI in human plasma by coagulometric methods.
Factor XI Deficient Plasma is a human plasma-based reagent for the detection of hereditary or acquired deficiencies of factor XI. The reagent has a residual factor concentration of <1% factor XI and was designed to be used in combination with Dade Actin, Dade Actin FS, Dade Actin FSL, or Pathromtin SL reagents.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Single Factor Testing
Quantity 3 x 1 mL
WARNING: Cancer - www.P65Warnings.ca.gov