Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor XI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547327
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547327 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547327 Supplier Invitrogen™ Supplier No. PA547327
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse F11 is observed and less than 5% cross-reactivity with recombinant human (rh) F7 and rhF10 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes coagulation factor XI of the blood coagulation cascade. This protein is present in plasma as a zymogen, which is a unique plasma coagulation enzyme because it exists as a homodimer consisting of two identical polypeptide chains linked by disulfide bonds. During activation of the plasma factor XI, an internal peptide bond is cleaved by factor XIIa (or XII) in each of the two chains, resulting in activated factor XIa, a serine protease composed of two heavy and two light chains held together by disulfide bonds. This activated plasma factor XI triggers the middle phase of the intrisic pathway of blood coagulation by activating factor IX. Defects in this factor lead to Rosenthal syndrome, a blood coagulation abnormality.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor XI
Applications Immunoprecipitation, Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene F11
Gene Accession No. P03951
Gene Alias 1600027G01Rik; AI503996; Cf11; Coagulation factor XI; Coagulation factor XIa heavy chain; Coagulation factor XIa light chain; F11; FXI; LRRGT00086; plasma thromboplastin antecedent; PTA
Gene Symbols F11
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor XI Glu19-Val625.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2160
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.