Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor XII Light chain Monoclonal Antibody (C6B7)
GREENER_CHOICE

Catalog No. PIMA126322
Change view
Click to view available options
Quantity:
125 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA126322 125 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA126322 Supplier Invitrogen™ Supplier No. MA126322
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes coagulation factor XII which circulates in blood as a zymogen. This single chain zymogen is converted to a two-chain serine protease with an heavy chain (alpha-factor XIIa) and a light chain. The heavy chain contains two fibronectin-type domains, two epidermal growth factor (EGF)-like domains, a kringle domain and a proline-rich domain, whereas the light chain contains only a catalytic domain. On activation, further cleavages takes place in the heavy chain, resulting in the production of beta-factor XIIa light chain and the alpha-factor XIIa light chain becomes beta-factor XIIa heavy chain. Prekallikrein is cleaved by factor XII to form kallikrein, which then cleaves factor XII first to alpha-factor XIIa and then to beta-factor XIIa. The active factor XIIa participates in the initiation of blood coagulation, fibrinolysis, and the generation of bradykinin and angiotensin. It activates coagulation factors VII and XI. Defects in this gene do not cause any clinical symptoms and the sole effect is that whole-blood clotting time is prolonged.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor XII Light chain
Applications ELISA, Western Blot
Classification Monoclonal
Clone C6B7
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene F12
Gene Accession No. P00748, Q80YC5
Gene Alias Beta-factor XIIa part 1; beta-factor XIIa part 2; Coagulation factor XII; coagulation factor XII (Hageman factor); coagulation factor XIIa heavy chain; Coagulation factor XIIa light chain; F12; Factor 12; Factor12; FXII; HAE3; HAEX; HAF; Hageman factor
Gene Symbols F12
Host Species Mouse
Immunogen Purified human Factor XII.
Purification Method Protein G
Quantity 125 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2161, 58992
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.