Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor XII Mouse anti-Baboon, Human, Clone: 15H8, MilliporeSigma™

Catalog No. p-7159477 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS2037-25 25 μg
MABS2037-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS2037-25 Supplier MilliporeSigma Supplier No. MABS203725UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Factor XII, clone 15H8 is a mouse monoclonal antibody that detects Coagulation factor XII and has been tested for use in ELISA, Inhibition and Function assays, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor XII
Applications ELISA, Inhibition Assays, Western Blot
Classification Monoclonal
Clone 15H8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols F12
Host Species Mouse
Immunogen A mixture of purified human Factor XII and Factor XIIa
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000496
Test Specificity Clone 15H8 is a mouse monoclonal antibody that specifically detects Coagulation factor XII.
Target Species Baboon, Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.