Learn More
Invitrogen™ Factor XII Polyclonal Antibody

Description
PA1-43049 detects Factor XII in human samples. PA1-43049 has successfully been used in Western blot and ELISA applications. PA1-43049 was derived from human factor XII.
Specifications
Specifications
| Antigen | Factor XII |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 8.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene | F12 |
| Gene Accession No. | P00748 |
| Gene Alias | beta-factor XIIa part 1; Beta-factor XIIa part 2; coagulation factor XII; coagulation factor XII (Hageman factor); coagulation factor XIIa heavy chain; Coagulation factor XIIa light chain; F12; F13; Factor 12; factor XII; Factor12; FXII; HAE3; HAEX; HAF; hageman factor |
| Gene Symbols | F12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.