Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Factor XIII Recombinant Rabbit Monoclonal Antibody (K01_1K61)
GREENER_CHOICE

Catalog No. MA563314
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA563314 100 μL
MA564773 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA563314 Supplier Invitrogen™ Supplier No. MA563314
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Factor XIII Recombinant Monoclonal Antibody for IHC (P), Western Blot

This gene encodes the coagulation factor XIII A subunit. Coagulation factor XIII is the last zymogen to become activated in the blood coagulation cascade. Plasma factor XIII is a heterotetramer composed of 2 A subunits and 2 B subunits. The A subunits have catalytic function, and the B subunits do not have enzymatic activity and may serve as plasma carrier molecules. Platelet factor XIII is comprised only of 2 A subunits, which are identical to those of plasma origin. Upon cleavage of the activation peptide by thrombin and in the presence of calcium ion, the plasma factor XIII dissociates its B subunits and yields the same active enzyme, factor XIIIa, as platelet factor XIII. This enzyme acts as a transglutaminase to catalyze the formation of gamma-glutamyl-epsilon-lysine crosslinking between fibrin molecules, thus stabilizing the fibrin clot. It also crosslinks alpha-2-plasmin inhibitor, or fibronectin, to the alpha chains of fibrin. Factor XIII deficiency is classified into two categories: type I deficiency, characterized by the lack of both the A and B subunits; and type II deficiency, characterized by the lack of the A subunit alone. These defects can result in a lifelong bleeding tendency, defective wound healing, and habitual abortion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor XIII
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1K61
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene F13A1
Gene Accession No. P00488
Gene Alias bA525O21.1 (coagulation factor XIII, A1 polypeptide); coagulation factor XIII; coagulation factor XIII A chain; Coagulation factor XIII B chain; coagulation factor XIII, A polypeptide; coagulation factor XIII, A1 polypeptide; coagulation factor XIII, B polypeptide; coagulation factor XIIIa; F13; F13A; F13A1; F13B; Factor 13; factor viiia; factor XIIIa; Factor13; fibrin stabilizing factor, A subunit; fibrinoligase; fibrin-stabilizing factor B subunit; FSF, A subunit; FXIIIB; Protein-glutamine gamma-glutamyltransferase A chain; protein-glutamine gamma-glutamyltransferase B chain; TGase; transglutaminase A chain; Transglutaminase B chain; transglutaminase. plasma
Gene Symbols F13A1
Host Species Rabbit
Immunogen Recombinant protein of human Factor XIIIa.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2162
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.