Learn More
Diagnostic Biosystems FACTOR XIIIA Factor XIII-A
Supplier:  Diagnostic Biosystems RP103
Encompass_Preferred
This antibody reacts with a 160 kDa (unreacted dimer) and 80 kDa (reduced monomer) protein. Factor XIII is a β-globulin found in plasma and is composed of two subunits, Factor XIII-A and Factor XIII-B. Factor XIII-A is the catalytic subunit and is a dimer of M.W. 160 kDa. Factor XIII is a dermal dendrocyte marker and shows variable reaction with these types of tumors. Factor XIII can be used for histiocytic phenotyping and has been reported to mark capillary hemangiomas and tumors of the central nervous system.
Catalog No. 50-238-5400
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.