Learn More
Diagnostic Biosystems FACTOR XIIIA Factor XIII-A (AC-1A1)
Supplier:  Diagnostic Biosystems MOB32105
Encompass_Preferred
This antibody is specific to 160 kDa protein known as Factor XIII-A. Factor XIII is a β-globulin found in plasma and is composed of two subunits, Factor XIIIA and Factor XIII-B. Factor XIII-A is the catalytic subunit and is a dimer of M.W. 160 kDa. Factor XIII is a dermal dendrocyte marker and v variable reaction with these types of tumors.
Catalog No. 50-238-5779
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.