Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAF1 Mouse anti-Human, Unconjugated, Clone: CPTC-FAF1-2, Abnova™

Catalog No. 89337640 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-640 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-640 Supplier Abnova Corporation Supplier No. MAB21147
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

FAF1 Monoclonal antibody specifically detects FAF1 in Human samples. It is validated for WB, IHC-P.

Specifications

Antigen FAF1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-FAF1-2
Conjugate Unconjugated
Description Fas (TNFRSF6) associated factor 1
Gene FAF1
Gene Alias CGI-03, FLJ37524, HFAF1s, UBXD12, UBXN3A, hFAF1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human FAF1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 11124
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.