Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529330 Supplier Invitrogen™ Supplier No. PA529330
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (98%), Rat (97%), Zebrafish (84%), Xenopus laevis (84%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Interaction of Fas ligand (TNFSF6) with the FAS antigen (TNFRSF6) mediates programmed cell death, also called apoptosis, in a number of organ systems. The protein encoded by this gene binds to FAS antigen and can initiate apoptosis or enhance apoptosis initiated through FAS antigen. Initiation of apoptosis by the protein encoded by this gene requires a ubiquitin-like domain but not the FAS-binding domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAF1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene FAF1
Gene Accession No. Q9UNN5
Gene Alias AA408698; CGI-03; Dffrx; Faf1; Fam; Fas (TNFRSF6) associated factor 1; Fas associated factor 1; Fas-associated factor 1; hFAF1; HFAF1s; TNFRSF6-associated factor 1; UBX domain protein 3A; UBX domain-containing protein 12; UBX domain-containing protein 3A; UBXD12; UBXN3A
Gene Symbols FAF1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FAF1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11124
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.