Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596220
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596220 Supplier Invitrogen™ Supplier No. PA596220
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASNMDREMI LADFQACTGI ENIDEAITLL EQNNWDLVAA INGVIPQENG ILQSEYGGET IPGPAFNPAS HPASAPTSSS SSAFRPVMPS RQIVERQPRM LDFRVEYRDR NVDVVLEDTC TVGEIKQILE NELQIPVSKM LLKGWKTGDV EDSTVLKSLH LPKNNSLYVL TPDLPPPSSS SHAGALQESL NQNFMLIITH REVQREYNLN FSGSSTIQEV KRNVYDLTSI PVRHQLWEGW PTSATDDSMC LAESGLSYPC HRLTVGRRSS PAQTREQSEE; Positive Samples: OVCAR3, HT-29, Mouse brain, Mouse liver, Rat brain; Cellular Location: Nucleus.

Interaction of Fas ligand (TNFSF6) with the FAS antigen (TNFRSF6) mediates programmed cell death, also called apoptosis, in a number of organ systems. The protein encoded by this gene binds to FAS antigen and can initiate apoptosis or enhance apoptosis initiated through FAS antigen. Initiation of apoptosis by the protein encoded by this gene requires a ubiquitin-like domain but not the FAS-binding domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAF1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FAF1
Gene Accession No. P54731, Q924K2, Q9UNN5
Gene Alias AA408698; CGI-03; Dffrx; Faf1; Fam; Fas (TNFRSF6) associated factor 1; Fas associated factor 1; Fas-associated factor 1; hFAF1; HFAF1s; TNFRSF6-associated factor 1; UBX domain protein 3A; UBX domain-containing protein 12; UBX domain-containing protein 3A; UBXD12; UBXN3A
Gene Symbols FAF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human FAF1 (NP_0089821).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11124, 140657, 14084
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.