Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Falcon™ Sterile Polyethylene Snap Caps
BIOPHARMAGSA_VAGREENER_CHOICE

Catalog No. 1495948 Shop All Corning Products
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-959-48 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-959-48 Supplier Falcon™ Supplier No. 352032
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Falcon Round-Bottom Tubes

These polyethylene caps feature a patented, two-position fit; loose for aerobic culturing, tight for sealing.

Falcon™ polyethylene dual-position snap caps for 12x75mm tubes offer both vented and fully closed positions. These sterile caps are packaged 500 per bag and 2,000 per case. RNase/DNase-free.
  • Dual position snap cap, offering vented as well as fully closed options
  • Sterilized by gamma irradiation
  • For use with both polystyrene and polypropylene tubes
  • Order Info

    Supplied in bags of 500; case of four bags.

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    Product Type Snap Cap
    Autoclavable Not Autoclavable
    Material Polyethylene
    Color Natural
    No. per Pack 500/Pk.
    No. per Case 2000/Cs.
    Sterility Sterile
    For Use With (Equipment) Falcon™ 75 L x 12 mm O.D. 5 mL polystyrene and polypropylene round bottom tubes
    Disposable Single-use
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.