Learn More
BD Falcon™ Swube™ Applicators
Catalog No. 1495975 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Product Type:
Single Swab attached to Screw Cap, Plastic Tube
Single Swabs Only
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
14-959-75 Single Swab attached to Screw Cap, Plastic Tube
14-959-85 Single Swabs Only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-959-75 Supplier BD Supplier No. 220710
This item is not returnable. View return policy
Sterile, disposable swabs
220710: Consists of a polyester-tipped applicator, approximately 15.2 cm (6”) in length, attached to the screw cap of a 16 x 150mm clear plastic tube.
  • Compact applicators
  • Collect specimens and protect them from contamination
  • Tested for microbial compatibility and sterility
Order Info
Individually wrapped
TRUSTED_SUSTAINABILITY
Specifications
Applicator Material Wood
Tip Material Polyester
Includes Polyester-tipped applicator, Clear plastic tube
Sterility Sterile
Packaging Individually-wrapped
For Use With (Application) For specimen collection and transportation
Length (English) 16 x 150
Product Type Single Swab attached to Screw Cap, Plastic Tube