Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific False Bottom Analyzer Tube
GSA_VA

Catalog No. 14375439 Shop All Starplex Scientific Products
Change view
Click to view available options
Color:
Clear
White
Product Type:
False Bottom Analyzer Vial
Screw Cap
Material:
Plastic
Polypropylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Material
14-375-439 Clear False Bottom Analyzer Vial Polypropylene
14-375-441 White Screw Cap Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-375-439 Supplier Starplex™ Scientific Supplier No. V8991
Add to Cart
Add to Cart

Designed for high-volume analyzer aspiration probes incapable of reaching the bottoms of larger sample tubes

  • Ideal for primary or secondary sampling as well as storage
  • No need for micro sample cups and liquid transfer devices
  • Accept bar codes and fit into most high volume analyzer racks
  • Compatible with Hitachi™, Olympus™, Corning™, Ortho (Kodak™), VITROS™ (750 and 250 series), and Beckman™ clinical chemistry analyzer

Order Info

Plug caps sold separately.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Material Polypropylene
Product Type False Bottom Analyzer Vial
Color Clear
Sterility Non-sterile
Volume (Metric) 5 mL
For Use With (Application) Designed for high-volume analyzer aspiration probes incapable of reaching the bottoms of larger sample tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.