Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fam Benzine, Solstice

Catalog No. 60048446 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-446 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-446 Supplier Solstice Supplier No. 245382.5L
Add to Cart
Add to Cart

DIN 51635, bp 65-95 °C

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Density 0.690 to 0.705g/mL at 15°C
Flash Point −26°C (−14.8°F)
Quantity 2.5 L
UN Number 1268
Chemical Name or Material Fam Benzine
Hazard Category Danger
Hazard Statement P201-P210-P280-P308 + P313-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H340-H350-H361f-H373-H412
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.