Learn More
CPC Scientific 5FAM-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-OH (trifluoroacetate salt) 0.1MG
SDP
Supplier:  CPC Scientific AMYD025A
Encompass
Synthetic peptide that forms fibrils with have the same antigenic and structural features as those of native AD amyloid filaments.ONE-LETTER SEQUENCE: 5FAM-DAEFRHDSGYEVHHQKLVFFAEDVGSNKMOLECULAR FORMULA: C166H219N41O52MOLECULAR WEIGHT:3620.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [109770-29-8], RESEARCH AREA: Alzheimer's DiseaseREFERENCES: J. Kang et al., Nature, 325, 773 (1987)
Catalog No. 50-210-8678
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.