Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FAM-DEALAHypYIPMDDDFQLRSF

Catalog No. 74521
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-521 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 74-521 Supplier Tocris Bioscience™ Supplier No. 7452/1
This item is not returnable. View return policy

Fluorescent HIF-1α peptide; can be used to evaluate the effect of VHL binding on degradation activity

FAM-DEALAHypYIPMDDDFQLRSF is a 5,6-carboxyfluorescein (FAM) labeled HIF-1α peptide. It binds with high affinity to von Hippel-Lindau (VHL) protein (KD = 3 nM). Can be used to assess ligand binding to VHL in a direct fluorescence polarization (FP) displacement assay. Excitation/emission maxima (λ) = 485/520 nm. This product is a longer peptide version of FAM-DEALA-Hyp-YIPD (Cat. No. 7287).

Specifications

Purity 95%
Content And Storage Store at -20°C
Molecular Weight (g/mol) M.W.-observed: 2616.76
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.