Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM103A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7209212 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2396 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2396 Supplier Proteintech Group Inc Supplier No. 194221AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM103A1, also named as RNMT-activating mini protein, is a 118 amino acid protein, which belongs to the RAM family. FAM103A1 as a component of mRNA cap binding complex localizes in the nucleus. FAM103A1 is required for efficient mRNA cap methylation and regulates RNMT expression by a post-transcriptional stabilizing mechanism.

Specifications

Antigen FAM103A1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FAM103A1
Gene Accession No. Q9BTL3, Q9CQY2
Gene Alias C15orf18, FAM103A1, HsT19360, Protein FAM103A1
Gene Symbols Fam103a1
Host Species Rabbit
Immunogen FAM103A1 Fusion Protein Ag13749
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293058, 67148, 83640
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.