Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM110C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557543 Supplier Invitrogen™ Supplier No. PA557543
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDPEVVEALG RENFTAGSDC VTLKVRSVSV ATSGSGFSRH SGGDDEGLQE EELIEQVPST TSVIE Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 77%.

FAM110C may play a role in microtubule organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM110C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM110C
Gene Accession No. Q1W6H9
Gene Alias FAM110C; family with sequence similarity 110 member C; family with sequence similarity 110, member C; protein FAM110C
Gene Symbols FAM110C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM110C. Recombinant protein control fragment (Product #RP-102699).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 642273
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.